missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 0.0100 N (N/100), Ricca Chemical
$117.13 - $117.13
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | HNO3 |
| Molecular Weight (g/mol) | 63.01 |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
| PubChem CID | 944 |
| ChEBI | CHEBI:48107 |
| IUPAC Name | nitric acid |
| SMILES | O[N+]([O-])=O |
Chemical Identifiers
| 7732-18-5 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
Specifications
| 7732-18-5,7697-37-2 | |
| 1 L | |
| MFCD00011349 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| Laboratory |
| Natural Poly Bottle | |
| HNO3 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| O[N+]([O-])=O | |
| 63.01 | |
| CHEBI:48107 | |
| Nitric acid |