missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrate Standard, 1000 ppm NO3- (226 ppm N), Ricca Chemical
$239.00 - $239.00
Chemical Identifiers
| CAS | 7757-79-1 |
|---|---|
| Molecular Formula | KNO3 |
| Molecular Weight (g/mol) | 101.10 |
| MDL Number | MFCD00011409 |
| InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
| PubChem CID | 24434 |
| ChEBI | CHEBI:63043 |
| IUPAC Name | potassium nitrate |
| SMILES | [K+].[O-][N+]([O-])=O |
Chemical Identifiers
| 7757-79-1 | |
| 101.10 | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| CHEBI:63043 | |
| [K+].[O-][N+]([O-])=O |
| KNO3 | |
| MFCD00011409 | |
| 24434 | |
| potassium nitrate |
Specifications
| Natural Poly Bottle | |
| 98.0,0.16 | |
| KNO3 | |
| 100°C | |
| [K+].[O-][N+]([O-])=O | |
| 101.10 | |
| 24434 | |
| Nitrate Standard, 1 mL = 1 mg NO3 |
| 7757-79-1,7732-18-5 | |
| 100.0,0.17 | |
| MFCD00011409 | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| potassium nitrate | |
| 16 oz. | |
| CHEBI:63043 |