missing translation for 'onlineSavingsMsg'
Learn More

Nitrate Nitrogen Standard, 1000 ppm N (4427 ppm NO3-), Ricca Chemical

$84.50 - $204.78

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5459-4
View Documents
Ricca Chemical Company
54594
120 mL Natural Poly Bottle
Each for $84.50
Only null left
 
5459-16
SDS View Documents
Ricca Chemical Company
545916
500 mL Natural Poly Bottle
Each for $204.78
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7757-79-1
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
CHEBI:63043
[K+].[O-][N+]([O-])=O
KNO3
MFCD00011409
24434
potassium nitrate

Specifications

Natural Poly Bottle
97.0,0.7
ASTM/EPA for Nitrate Nitrogen
KNO3
100°C
[K+].[O-][N+]([O-])=O
101.10
24434
1000ppm N as nitrate, 4427ppm NO3-
7757-79-1,7732-18-5
100.0,0.73
1mL = 1mgN
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate
120 mL
CHEBI:63043
Nitrogen Standard (as Nitrate)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'