missing translation for 'onlineSavingsMsg'
Learn More

Nitinol tension coil spring, Alloy M, Thermo Scientific™

Catalog No. AA45655KN
Change view
Click to view available options
Quantity:
100 pc.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45655KN 100 pc.
AA45655KM -
AA45655KJ -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA45655KN Supplier Thermo Scientific Chemicals Supplier No. 45655 KN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7440-02-0 , 7440-32-6
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 100 pc.
Odor Odorless
Concentration or Composition (by Analyte or Components) Nickel: 55.75%; Titanium: 44.25%
Chemical Name or Material Nitinol tension coil spring, Wire diameter 0.75mm, mean coil diameter 5.55mm
Hazard Category H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only