missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Nitinol-core (NIT) SPME Fiber Assembly Carboxen/Polydimethylsiloxane (Carboxen/PDMS)
For Gases and Low Molecular Weight Compounds (MW 30-225)
$1,133.23
Specifications
| Film Thickness | 75 μm |
|---|---|
| Material | Black Hub (Metallic) |
| Matrix | Carboxen/Polydimethylsiloxane (CAR/PDMS) coating |
| Quantity | 3/Pk. |
| Catalog Number | Mfr. No. | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
|
111021704
|
MilliporeSigma™ Supelco™
57901U |
Manual Holder | Needle Size of 23 ga |
Each for $1,133.23
|
|
||||
|
111021705
|
MilliporeSigma™ Supelco™
57904U |
Manual Holder | Needle Size of 24 ga |
Each for $1,133.23
|
|
||||
|
111021706
|
MilliporeSigma™ Supelco™
57907U |
Autosampler or Manual Holder | Needle Size of 23 ga |
Each for $1,133.23
|
|
||||
|
111021707
|
MilliporeSigma™ Supelco™
57908U |
Autosampler or Manual Holder | Needle Size of 24 ga |
Each for $1,133.23
|
|
||||
Description
- Nitinol-core (NIT) SPME fibers were specifically engineered to be more reproducible and more robust than traditional SPME fibers that contain a fused silica (FS) core.
- The NIT fibers offer > 30% improvement in lot to lot and fiber to fiber reproducibility when compared to the FS versions, but offer equivalent selectivity for easy transfer of methods from FS to the more robust, reproducible NIT version.
Specifications
| 75 μm | |
| Black Hub (Metallic) | |
| Carboxen/Polydimethylsiloxane (CAR/PDMS) coating | |
| 3/Pk. |