missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Nitinol-core (NIT) SPME Fiber Assembly Carboxen/Polydimethylsiloxane (Carboxen/PDMS)

Catalog No. 111021704
Change view
Click to view available options
For Use With (Equipment):
Autosampler or Manual Holder
Manual Holder
Includes:
Needle Size of 23 ga
Needle Size of 24 ga
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Includes
111021704 Manual Holder Needle Size of 23 ga
111021705 Manual Holder Needle Size of 24 ga
111021706 Autosampler or Manual Holder Needle Size of 23 ga
111021707 Autosampler or Manual Holder Needle Size of 24 ga
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111021704 Supplier MilliporeSigma™ Supelco™ Supplier No. 57901U

For Gases and Low Molecular Weight Compounds (MW 30-225)

  • Nitinol-core (NIT) SPME fibers were specifically engineered to be more reproducible and more robust than traditional SPME fibers that contain a fused silica (FS) core.
  • The NIT fibers offer > 30% improvement in lot to lot and fiber to fiber reproducibility when compared to the FS versions, but offer equivalent selectivity for easy transfer of methods from FS to the more robust, reproducible NIT version.

Specifications

For Use With (Equipment) Manual Holder
Film Thickness 75 μm
Includes Needle Size of 23 ga
Material Black Hub (Metallic)
Matrix Carboxen/Polydimethylsiloxane (CAR/PDMS) coating
Quantity 3/Pk.