missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Nitarsone | |
| 300°C | |
| C6H6AsNO5 | |
| 1 g | |
| 4-Nitrophenylarsonic Acid, 4-Nitrobenzenearsonic Acid | |
| O[As](O)(=O)C1=CC=C(C=C1)[N+]([O-])=O | |
| 247.04 | |
| CHEBI:62629 | |
| ≥98.0% (HPLC,T) |
| 98-72-6 | |
| Yellow | |
| MFCD00024575 | |
| 3465 | |
| FUUFQLXAIUOWML-UHFFFAOYSA-N | |
| (4-nitrophenyl)arsonic acid | |
| 66826 | |
| 247.04 | |
| Crystalline Powder |
Chemical Identifiers
| 98-72-6 | |
| 247.04 | |
| FUUFQLXAIUOWML-UHFFFAOYSA-N | |
| 66826 | |
| (4-nitrophenyl)arsonic acid |
| C6H6AsNO5 | |
| MFCD00024575 | |
| 4-Nitrophenylarsonic Acid, 4-Nitrobenzenearsonic Acid | |
| CHEBI:62629 | |
| O[As](O)(=O)C1=CC=C(C=C1)[N+]([O-])=O |
Safety and Handling
TSCA : No