Learn More
Beckman Coulter NIST Traceable Particle Size Controls
Particle Size Controls for use with LS 13 320 XR Analyzers to verify instrument and module performance.
Supplier: Beckman Coulter C12537
Description
- For verifying instrument and module performance
- Recommended for LS 13 320 XR instruments
- NIST traceable
Chemical Identifiers
| Particle Size Control | |
| Verifying Instrument and Module Perfomance | |
| LS 13 320 XR Analyzer |
Specifications
| Particle Size Control | |
| Polystyrene | |
| Verifying Instrument and Module Perfomance |
| 15 mL | |
| 0.04 μm | |
| LS 13 320 XR Analyzer |