missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets

Description
pH measurements are only as accurate as the buffer solutions used for calibration purposes. Maximum precision is guaranteed with fully traceable NIST/DIN buffers. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)
- NIST/DIN pH buffers are specified and guaranteed to three decimal places, offering top precision
- Individual sachets guarantee fresh solution for every calibration, as well as maximum ease of use
- A test certificate and detailed label with a temperature table comes with every box
- All SDS and labels contain information according to GHS (Globally Harmonized System)
Specifications
Specifications
| Description | pH Buffer |
| For Use With (Application) | pH measurement, electrode calibration |
| For Use With (Equipment) | pH Electrode |
| pH | 10.012 (at 25°C) |
| Quantity | 30 x 20 mL |
| Packaging | 20 mL Sachets |
| Traceability to NIST | Yes |