Learn More
Thermco™ NIST Certification
For use with HITEMP135Autoclave Temperature Data Loggers
Supplier: Thermco™ NISTHITEMP
Description
- Makes HITEMP135 data loggers traceable to N.I.S.T. to comply with regulatory agencies
- N. I. S. T. ISO17025/ILAC Certification certificate for HITEMP135 data loggers, 1 point
Specifications
| NIST Certification | |
| HITEMP135 Autoclave Temperature Data Loggers |