Learn More
Niobium(V) fluoride, 99%
CAS: 7783-68-8 | F5Nb | 187.90 g/mol
$243.62 - $942.67
Chemical Identifiers
| CAS | 7783-68-8 |
|---|---|
| Molecular Formula | F5Nb |
| Molecular Weight (g/mol) | 187.90 |
| MDL Number | MFCD00011129 |
| InChI Key | AOLPZAHRYHXPLR-UHFFFAOYSA-I |
| Synonym | Niobium pentafluoride |
| IUPAC Name | niobium(5+) pentafluoride |
| SMILES | [F-].[F-].[F-].[F-].[F-].[Nb+5] |
Description
Niobium(V) fluoride is used as a starting material to prepare other niobium compounds. It acts as an intermediate during the recovery of niobium metal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7783-68-8 | |
| 187.90 | |
| AOLPZAHRYHXPLR-UHFFFAOYSA-I | |
| niobium(5+) pentafluoride |
| F5Nb | |
| MFCD00011129 | |
| Niobium pentafluoride | |
| [F-].[F-].[F-].[F-].[F-].[Nb+5] |
Specifications
| ∼230°C | |
| 7783-68-8 | |
| Powder | |
| F5Nb | |
| UN3260 | |
| Niobium pentafluoride | |
| AOLPZAHRYHXPLR-UHFFFAOYSA-I | |
| niobium(5+) pentafluoride | |
| 187.91 | |
| Moisture sensitive | |
| Niobium (V) fluoride |
| 80°C | |
| White to Yellow | |
| 10 g | |
| MFCD00011129 | |
| 14,6558 | |
| Hydrolyzes in water,alcohol,and caustic solutions; Sparingly soluble in CS2,chloroform; Forms complexes with Lewis basesSlightly soluble in carbon disulfide and chloroform. | |
| [F-].[F-].[F-].[F-].[F-].[Nb+5] | |
| 187.90 | |
| 99% | |
| 3.92 g/mL |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 232-020-2
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only