Learn More
Niobium n-butoxide, 99% (metals basis)
CAS: 51030-47-8 | C20H50NbO5 | 463.521 g/mol
$107.33 - $1188.05
Chemical Identifiers
| CAS | 51030-47-8 |
|---|---|
| Molecular Formula | C20H50NbO5 |
| Molecular Weight (g/mol) | 463.521 |
| MDL Number | MFCD00144916 |
| InChI Key | OMBQVKDAXPUDBD-UHFFFAOYSA-N |
| Synonym | butan-1-ol; niobium, niobium v-n-butoxide |
| PubChem CID | 50912030 |
| IUPAC Name | butan-1-ol;niobium |
| SMILES | CCCCO.CCCCO.CCCCO.CCCCO.CCCCO.[Nb] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA3657003
|
Thermo Scientific Chemicals
03657003 |
1 g |
Each for $107.33
|
|
|||||
|
AA3657006
|
Thermo Scientific Chemicals
03657006 |
5 g |
Each for $323.05
|
|
|||||
|
AA3657014
|
Thermo Scientific Chemicals
03657014 |
25 g |
Each for $1,188.05
|
|
|||||
Description
Niobium n-butoxide plays an important role as a precursor in chemical synthesis. It is also used in the production of nano-materials. It acts as a precursor for dielectric films and structures with controlled properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 51030-47-8 | |
| 463.521 | |
| OMBQVKDAXPUDBD-UHFFFAOYSA-N | |
| 50912030 | |
| CCCCO.CCCCO.CCCCO.CCCCO.CCCCO.[Nb] |
| C20H50NbO5 | |
| MFCD00144916 | |
| butan-1-ol; niobium, niobium v-n-butoxide | |
| butan-1-ol;niobium |
Specifications
| 51030-47-8 | |
| 197°C (0.15 mmHg) | |
| Nb[O(CH2)3CH3]5 | |
| 1 g | |
| butan-1-ol; niobium, niobium v-n-butoxide | |
| OMBQVKDAXPUDBD-UHFFFAOYSA-N | |
| butan-1-ol;niobium | |
| 50912030 | |
| 99% | |
| Niobium n-butoxide |
| 1.09 g/mL | |
| C20H50NbO5 | |
| MFCD00144916 | |
| Moisture sensitive | |
| Miscible with hydrocarbons,ketones and esters. | |
| CCCCO.CCCCO.CCCCO.CCCCO.CCCCO.[Nb] | |
| 463.521 | |
| 458.12 | |
| Liquid | |
| (metals basis) |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
EINECSNumber : 256-923-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only