accessibility menu, dialog, popup

UserName

CAS RN 51030-47-8

NbH3COCH3OCH3OCH3OCH3O

CAS RN 51030-47-8

IUPAC Name: niobium(5+) pentakis(butan-1-olate)
Synonyms: niobium(5+) pentakis(butan-1-olate)
Molecular Weight (g/mol): 458.48
Molecular Formula: C20H45NbO5
InChi Key: DINQVNXOZUORJS-UHFFFAOYSA-N
SMILES: [Nb+5].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-]
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results