missing translation for 'onlineSavingsMsg'
Learn More

Niobium n-butoxide, 99% (metals basis)

Catalog No. AA3657003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3657003 1 g
AA3657006 5 g
AA3657014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3657003 Supplier Thermo Scientific Chemicals Supplier No. 03657003

CAS: 51030-47-8 | C20H50NbO5 | 463.521 g/mol

Niobium n-butoxide plays an important role as a precursor in chemical synthesis. It is also used in the production of nano-materials. It acts as a precursor for dielectric films and structures with controlled properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51030-47-8
Molecular Formula C20H50NbO5
Molecular Weight (g/mol) 463.521
MDL Number MFCD00144916
InChI Key OMBQVKDAXPUDBD-UHFFFAOYSA-N
Synonym butan-1-ol; niobium, niobium v-n-butoxide
PubChem CID 50912030
IUPAC Name butan-1-ol;niobium
SMILES CCCCO.CCCCO.CCCCO.CCCCO.CCCCO.[Nb]

Specifications

Density 1.09 g/mL
Boiling Point 197°C (0.15 mmHg)
Linear Formula Nb[O(CH2)3CH3]5
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Miscible with hydrocarbons,ketones and esters.
Formula Weight 458.12
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Niobium n-butoxide
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 256-923-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only