Learn More
Thermo Scientific Chemicals Ninhydrin, ACS reagent
CAS: 485-47-2 | C9H6O4 | 178.143 g/mol
$131.60 - $1115.96
Chemical Identifiers
| CAS | 485-47-2 |
|---|---|
| Molecular Formula | C9H6O4 |
| Molecular Weight (g/mol) | 178.143 |
| MDL Number | MFCD00003791 |
| InChI Key | FEMOMIGRRWSMCU-UHFFFAOYSA-N |
| Synonym | 2,2-Dihydroxy-1,3-indanedione |
| PubChem CID | 10236 |
| ChEBI | CHEBI:86374 |
| IUPAC Name | 2,2-dihydroxyindene-1,3-dione |
| SMILES | C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4384614
|
Thermo Scientific Chemicals
04384614 |
25 g |
Each for $131.60
|
|
|||||
|
AA4384622
|
Thermo Scientific Chemicals
04384622 |
100 g |
Each for $346.45
|
|
|||||
|
AA4384636
|
Thermo Scientific Chemicals
04384636 |
500 g |
Each for $1,115.96
|
|
|||||
Description
Reagent for determination of amino acids and amine containing compounds, as an intermediate.Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Reagent for determination of amino acids and amine containing compounds, as an intermediate
Chemical Identifiers
| 485-47-2 | |
| 178.143 | |
| FEMOMIGRRWSMCU-UHFFFAOYSA-N | |
| 10236 | |
| 2,2-dihydroxyindene-1,3-dione |
| C9H6O4 | |
| MFCD00003791 | |
| 2,2-Dihydroxy-1,3-indanedione | |
| CHEBI:86374 | |
| C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O |
Specifications
| 251°C (decomposition) | |
| C9H6O4 | |
| 1910963 | |
| 2,2-Dihydroxy-1,3-indanedione | |
| FEMOMIGRRWSMCU-UHFFFAOYSA-N | |
| 2,2-dihydroxyindene-1,3-dione | |
| 10236 | |
| 178.14 | |
| Light sensitive | |
| 25 g | |
| Ninhydrin |
| 485-47-2 | |
| MFCD00003791 | |
| 14,6554 | |
| Soluble in water,acetone,ethanol,and methanol; Slightly soluble in chloroform and ether. | |
| C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O | |
| 178.143 | |
| CHEBI:86374 | |
| ACS | |
| Brown to Yellow | |
| Crystalline |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 207-618-1
RTECSNumber : NK5425000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only