missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ninhydrin, ACS reagent

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol

Supplier:  Thermo Scientific Chemicals 04384622

 View more versions of this product

Catalog No. AA4384622


Only null left

Description

Description

Reagent for determination of amino acids and amine containing compounds, as an intermediate.Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Reagent for determination of amino acids and amine containing compounds, as an intermediate
Specifications

Specifications

Ninhydrin
485-47-2
MFCD00003791
14,6554
Soluble in water,acetone,ethanol,and methanol; Slightly soluble in chloroform and ether.
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
178.143
CHEBI:86374
ACS
Brown to Yellow
Crystalline
251°C (decomposition)
C9H6O4
1910963
2,2-Dihydroxy-1,3-indanedione
FEMOMIGRRWSMCU-UHFFFAOYSA-N
2,2-dihydroxyindene-1,3-dione
10236
178.14
Light sensitive
100 g

Chemical Identifiers

485-47-2
178.143
FEMOMIGRRWSMCU-UHFFFAOYSA-N
10236
2,2-dihydroxyindene-1,3-dione
C9H6O4
MFCD00003791
2,2-Dihydroxy-1,3-indanedione
CHEBI:86374
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 207-618-1

RTECSNumber : NK5425000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only