missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nimotuzumab Chimeric Recombinant Rabbit Monoclonal Antibody (h-R3 (Nimotuzumab))
GREENER_CHOICE

Catalog No. MA551603
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551603 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551603 Supplier Invitrogen™ Supplier No. MA551603
Only null left

Rabbit Recombinant Monoclonal Antibody

Nimotuzumab Chimeric Recombinant Monoclonal Antibody for Flow, ICC/IF, IHC, Inhibition

Nimotuzumab is a humanized therapeutic monoclonal antibody against epidermal growth factor receptor (EGFR).

Specifications

Antigen Nimotuzumab Chimeric
Applications Flow Cytometry, Inhibition Assays, Immunohistochemistry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone h-R3 (Nimotuzumab)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias hR3; Theraloc
Host Species Rabbit
Immunogen This antibody we generated by humanisation of the murine antibody R3, raised against EGFR purified from placenta membranes and A431 tumors.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less