missing translation for 'onlineSavingsMsg'
Learn More

Nimodipine, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 329280010

 View more versions of this product

Catalog No. AC329280010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Nimodipine
0.5% max. (105°C, 4 h)
C21H26N2O7
1g
nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate
UIAGMCDKSXEBJQ-UHFFFAOYNA-N
3-O-(2-methoxyethyl) 5-O-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
4497
418.44
66085-59-4
Authentic
MFCD00153848
15,6636
Solubility in water: insoluble. Other solubilities: soluble in dmso
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C
418.45
CHEBI:7575
≥97.5% (HPLC)

Chemical Identifiers

66085-59-4
418.45
UIAGMCDKSXEBJQ-UHFFFAOYNA-N
4497
3-O-(2-methoxyethyl) 5-O-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
C21H26N2O7
MFCD00153848
nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate
CHEBI:7575
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause damage to organs through prolonged or repeated exposure if swallowed.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical attention/advice if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 266-127-

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only