missing translation for 'onlineSavingsMsg'
Learn More

Nimodipine, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 329280050

Quantity 5g

 Afficher plus de versions de ce produit

Numéro de catalogue. AC329280050

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Identifiants chimiques

66085-59-4
418.45
UIAGMCDKSXEBJQ-UHFFFAOYNA-N
4497
3-O-(2-methoxyethyl) 5-O-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
C21H26N2O7
MFCD00153848
nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate
CHEBI:7575
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C

Spécifications

Nimodipine
0.5% max. (105°C, 4 h)
C21H26N2O7
5g
nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate
UIAGMCDKSXEBJQ-UHFFFAOYNA-N
3-O-(2-methoxyethyl) 5-O-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
4497
418.44
66085-59-4
Authentic
MFCD00153848
15,6636
Solubility in water: insoluble. Other solubilities: soluble in dmso
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C
418.45
CHEBI:7575
≥97.5% (HPLC)
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
May cause damage to organs through prolonged or repeated exposure if swallowed.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical attention/advice if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 266-127-

FDS
Documents

Documents