missing translation for 'onlineSavingsMsg'
Learn More

Nilvadipine 95.0+%, TCI America™

Catalog No. N08995G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08995G 5 g
N08991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08995G Supplier TCI America Supplier No. N08995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75530-68-6
Molecular Formula C19H19N3O6
Molecular Weight (g/mol) 385.38
MDL Number MFCD00867065
InChI Key FAIIFDPAEUKBEP-UHFFFAOYNA-N
Synonym 2-Cyano-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 5-Isopropyl 3-Methyl Ester, 5-Isopropyl 3-Methyl 2-Cyano-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate
PubChem CID 4494
IUPAC Name 3-methyl 5-propan-2-yl 2-cyano-6-methyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C)C#N

Specifications

Melting Point 151°C
Quantity 5 g
Formula Weight 385.38
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Nilvadipine
RTECSNumber US7968300
TSCA No