accessibility menu, dialog, popup

UserName

CAS RN 75530-68-6

IUPAC Name: 3-methyl 5-propan-2-yl 2-cyano-6-methyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: nilvadipine nivadil
Molecular Weight (g/mol): 385.38
Molecular Formula: C19H19N3O6
InChi Key: FAIIFDPAEUKBEP-UHFFFAOYNA-N
SMILES: COC(=O)C1=C(NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results