missing translation for 'onlineSavingsMsg'
Learn More

Nile Red, TCI America™

Catalog No. p-7133916
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0659500MG 500 mg
N06591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0659500MG Supplier TCI America Supplier No. N0659500MG
Only null left

Chemical Identifiers

CAS 7385-67-3
Molecular Formula C20H18N2O2
Molecular Weight (g/mol) 318.38
MDL Number MFCD00011639
InChI Key VOFUROIFQGPCGE-UHFFFAOYSA-N
Synonym 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
PubChem CID 65182
ChEBI CHEBI:52169
IUPAC Name 8-(diethylamino)-12H-10-oxa-5-azatetraphen-12-one
SMILES CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31

Specifications

Formula Weight 318.38
Color Green/Yellow
Quantity 500 mg
Physical Form Crystal/Powder
Chemical Name or Material Nile Red