missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nile Blue A, pure, certified

CAS: 3625-57-8 | C40H40N6O6S | 732.84 g/mol

$103.02 - $385.70

Chemical Identifiers

CAS 3625-57-8
Molecular Formula C40H40N6O6S
Molecular Weight (g/mol) 732.84
MDL Number MFCD00064529
InChI Key QIRDPEPUXNCOLD-UHFFFAOYSA-N
Synonym Nile blue sulfate, C.I. 51180
PubChem CID 19256
IUPAC Name [9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate
SMILES CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC415690100
SDS View Documents
Thermo Scientific Chemicals
415690100
10 g Glass bottle
Each for $103.02
Only null left
 
AC415690500
SDS View Documents
Thermo Scientific Chemicals
415690500
50 g Glass bottle
Each for $385.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3625-57-8
732.84
QIRDPEPUXNCOLD-UHFFFAOYSA-N
19256
CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]
C40H40N6O6S
MFCD00064529
Nile blue sulfate, C.I. 51180
[9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate

Specifications

3625-57-8
70% min.
MFCD00064529
Nile blue sulfate, C.I. 51180
Authentic
[9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate
19256
732.87
Glass bottle
10 g
Nile Blue A, Pure
1.01 to 1.16
C40H40N6O6S
27, 404
QIRDPEPUXNCOLD-UHFFFAOYSA-N
CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]
732.84
635 to 646nm
Pure
Black to Green
Powder
Videos
SDS
Documents

Documents

RUO – Research Use Only