missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Nigrosine, pure, water soluble
CAS: 8005-03-6 | C22H14N6Na2O9S2 | 616.49 g/mol
$132.57 - $322.00
Chemical Identifiers
| CAS | 8005-03-6 |
|---|---|
| Molecular Formula | C22H14N6Na2O9S2 |
| Molecular Weight (g/mol) | 616.49 |
| MDL Number | MFCD00044681 |
| InChI Key | HKBVRFLHNUEVRO-HMDKGIQMSA-L |
| Synonym | Acid Black 2, C.I. 50420 |
| PubChem CID | 44134531 |
| IUPAC Name | disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate |
| SMILES | C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC415681000
|
Thermo Scientific Chemicals
415681000 |
100 g | Glass bottle |
Each for $132.57
|
|
||||
|
AC415685000
|
Thermo Scientific Chemicals
415685000 |
500 g | Glass bottle |
Each for $322.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 8005-03-6 | |
| 616.49 | |
| HKBVRFLHNUEVRO-HMDKGIQMSA-L | |
| 44134531 | |
| C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
| C22H14N6Na2O9S2 | |
| MFCD00044681 | |
| Acid Black 2, C.I. 50420 | |
| disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate |
Specifications
| 8005-03-6 | |
| MFCD00044681 | |
| Solubility in water: soluble. | |
| C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] | |
| disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate | |
| 44134531 | |
| Complies | |
| Crystalline Powder or Crystals |
| C22H14N6Na2O9S2 | |
| Acid Black 2, C.I. 50420 | |
| HKBVRFLHNUEVRO-HMDKGIQMSA-L | |
| Authentic | |
| 616.49 | |
| Pure | |
| Black to Black/Red | |
| Nigrosine, pract. |
Safety and Handling
TSCA : TSCA
RUO – Research Use Only