missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Nifedipine | |
| 174°C | |
| C17H18N2O6 | |
| 10 g | |
| HYIMSNHJOBLJNT-UHFFFAOYSA-N | |
| 3,5-dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate | |
| 4485 | |
| 346.34 | |
| Crystalline Powder |
| 21829-25-4 | |
| Yellow | |
| MFCD00057326 | |
| nifedipine, procardia, adalat, procardia xl, adalat cc, cordipin, corinfar, fenihidine, citilat, oxcord | |
| COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OC | |
| 346.34 | |
| CHEBI:7565 | |
| ≥98.0% (HPLC,E) |
Chemical Identifiers
| 21829-25-4 | |
| 346.34 | |
| HYIMSNHJOBLJNT-UHFFFAOYSA-N | |
| 4485 | |
| 3,5-dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
| C17H18N2O6 | |
| MFCD00057326 | |
| nifedipine, procardia, adalat, procardia xl, adalat cc, cordipin, corinfar, fenihidine, citilat, oxcord | |
| CHEBI:7565 | |
| COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OC |
Safety and Handling
RTECSNumber : US7975000
TSCA : No