missing translation for 'onlineSavingsMsg'
Learn More

Nifedipine 98.0+%, TCI America™

Catalog No. p-7133861
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N052810G 10 g
N052825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N052810G Supplier TCI America Supplier No. N052810G

Chemical Identifiers

CAS 21829-25-4
Molecular Formula C17H18N2O6
Molecular Weight (g/mol) 346.34
MDL Number MFCD00057326
InChI Key HYIMSNHJOBLJNT-UHFFFAOYSA-N
Synonym nifedipine, procardia, adalat, procardia xl, adalat cc, cordipin, corinfar, fenihidine, citilat, oxcord
PubChem CID 4485
ChEBI CHEBI:7565
IUPAC Name 3,5-dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OC

Specifications

Melting Point 174°C
Color Yellow
Quantity 10 g
Formula Weight 346.34
Percent Purity ≥98.0% (HPLC,E)
Physical Form Crystalline Powder
Chemical Name or Material Nifedipine
RTECSNumber US7975000
TSCA No