missing translation for 'onlineSavingsMsg'
Learn More

Nicotinuric acid, 98+%, Thermo Scientific Chemicals

Catalog No. p-7058217
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6221506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6221506 Supplier Thermo Scientific Chemicals Supplier No. J6221506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 583-08-4 | Molecular Formula: C8H8N2O3 | Molecular Weight: 180.163 g/mol

Measurement of NUA levels in urine is used to indicate possible elevated nicotinamide adenine dinucleotide (NAD) catabolism. NUA may be used as a reference (standard curve) in analysis of NAD and NUA catabolism. It is an antiproliferative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Measurement of NUA levels in urine is used to indicate possible elevated nicotinamide adenine dinucleotide (NAD) catabolism. NUA may be used as a reference (standard curve) in analysis of NAD and NUA catabolism. It is an antiproliferative.

Solubility
Soluble in dimethyl sulfoxide, methanol, water, and 1M NH4OH (50 mg/mL).

Notes
Store at -20°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 583-08-4
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00023578
InChI Key ZBSGKPYXQINNGF-UHFFFAOYSA-N
Synonym nicotinuric acid, nicotinylglycine, nicotinoylglycine, 2-nicotinamido acetic acid, n-nicotinoylglycine, n-pyridin-3-ylcarbonyl glycine, nicotinoyl-glycine, n-nicotinylglycine, nicotinurate, n-nicotinoyl-glycine
PubChem CID 68499
ChEBI CHEBI:7563
IUPAC Name 2-(pyridine-3-carbonylamino)acetic acid
SMILES C1=CC(=CN=C1)C(=O)NCC(=O)O

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in dimethyl sulfoxide,methanol,water,and 1M NH4OH (50mg/mL).
Formula Weight 180.16
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Nicotinuric acid
EINECSNumber 209-497-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only