accessibility menu, dialog, popup

UserName

HOONHON

CAS RN 583-08-4

IUPAC Name: 2-[(pyridin-3-yl)formamido]acetic acid
Synonyms: nicotinuric acid N-nicotinylglycine nicotinurate nicotinylglycine
Molecular Weight (g/mol): 180.16
Molecular Formula: C8H8N2O3
InChi Key: ZBSGKPYXQINNGF-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)C1=CC=CN=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)