missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinoyl Chloride Hydrochloride 95.0+%, TCI America™
$83.89 - $1224.68
Chemical Identifiers
| CAS | 20260-53-1 |
|---|---|
| Molecular Formula | C6H5Cl2NO |
| Molecular Weight (g/mol) | 178.012 |
| MDL Number | MFCD00012820 |
| InChI Key | MSYBLBLAMDYKKZ-UHFFFAOYSA-N |
| Synonym | nicotinoyl chloride hydrochloride, pyridine-3-carbonyl chloride hydrochloride, nicotinoyl chloride, hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride, 3-pyridinecarbonyl chloride hydrochloride, nicotinic acid chloride hydrochloride, nicotinoylchloride hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride 1:1, nicotinyl chloride hydrochloride, nicotinoyl chloride hcl |
| PubChem CID | 88438 |
| IUPAC Name | pyridine-3-carbonyl chloride;hydrochloride |
| SMILES | C1=CC(=CN=C1)C(=O)Cl.Cl |
Chemical Identifiers
| 20260-53-1 | |
| 178.012 | |
| MSYBLBLAMDYKKZ-UHFFFAOYSA-N | |
| 88438 | |
| C1=CC(=CN=C1)C(=O)Cl.Cl |
| C6H5Cl2NO | |
| MFCD00012820 | |
| nicotinoyl chloride hydrochloride, pyridine-3-carbonyl chloride hydrochloride, nicotinoyl chloride, hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride, 3-pyridinecarbonyl chloride hydrochloride, nicotinic acid chloride hydrochloride, nicotinoylchloride hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride 1:1, nicotinyl chloride hydrochloride, nicotinoyl chloride hcl | |
| pyridine-3-carbonyl chloride;hydrochloride |
Specifications
| 20260-53-1 | |
| White | |
| MFCD00012820 | |
| 3261 | |
| MSYBLBLAMDYKKZ-UHFFFAOYSA-N | |
| pyridine-3-carbonyl chloride;hydrochloride | |
| 88438 | |
| ≥95.0% (T) | |
| Nicotinoyl Chloride Hydrochloride |
| 155°C | |
| C6H5Cl2NO | |
| 25 g | |
| nicotinoyl chloride hydrochloride, pyridine-3-carbonyl chloride hydrochloride, nicotinoyl chloride, hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride, 3-pyridinecarbonyl chloride hydrochloride, nicotinic acid chloride hydrochloride, nicotinoylchloride hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride 1:1, nicotinyl chloride hydrochloride, nicotinoyl chloride hcl | |
| C1=CC(=CN=C1)C(=O)Cl.Cl | |
| 178.012 | |
| 178.01 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0735
TSCA : Yes