accessibility menu, dialog, popup

UserName

HClClON

CAS RN 20260-53-1

IUPAC Name: hydrogen pyridine-3-carbonyl chloride chloride
Synonyms: hydrogen pyridine-3-carbonyl chloride chloride
Molecular Weight (g/mol): 178.01
Molecular Formula: C6H5Cl2NO
InChi Key: MSYBLBLAMDYKKZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC(=O)C1=CC=CN=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)