missing translation for 'onlineSavingsMsg'
Learn More

Nicotinoyl Chloride Hydrochloride 95.0+%, TCI America™

Catalog No. N009125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N009125G 25 g
N0091100G 100 g
N0091500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N009125G Supplier TCI America Supplier No. N009125G

Chemical Identifiers

CAS 20260-53-1
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.012
MDL Number MFCD00012820
InChI Key MSYBLBLAMDYKKZ-UHFFFAOYSA-N
Synonym nicotinoyl chloride hydrochloride, pyridine-3-carbonyl chloride hydrochloride, nicotinoyl chloride, hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride, 3-pyridinecarbonyl chloride hydrochloride, nicotinic acid chloride hydrochloride, nicotinoylchloride hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride 1:1, nicotinyl chloride hydrochloride, nicotinoyl chloride hcl
PubChem CID 88438
IUPAC Name pyridine-3-carbonyl chloride;hydrochloride
SMILES C1=CC(=CN=C1)C(=O)Cl.Cl

Specifications

Melting Point 155°C
Color White
Quantity 25 g
UN Number 3261
Formula Weight 178.01
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nicotinoyl Chloride Hydrochloride
EINECSNumber (5)-0735
TSCA Yes