Learn More
Nicotinoyl chloride hydrochloride, 96%
CAS: 20260-53-1 | C6H5Cl2NO | 178.012 g/mol
Supplier: Thermo Scientific Chemicals A1531718
Description
Nicotinoyl chloride hydrochloride was used in the preparation of 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil, bis(ethylenedithio)tetrathiafulvalene derivatives and 7,16-dinicotinoylated or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNicotinoyl chloride hydrochloride was used in the preparation of 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil, bis(ethylenedithio)tetrathiafulvalene derivatives and 7,16-dinicotinoylated or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes.
Solubility
It decomposes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications
| Nicotinoyl chloride hydrochloride | |
| 149°C to 155°C | |
| MFCD00012820 | |
| UN3261 | |
| Moisture sensitive | |
| It decomposes in water. | |
| C1=CC(=CN=C1)C(=O)Cl.Cl | |
| 178.012 | |
| 178.02 |
| 20260-53-1 | |
| C6H5Cl2NO | |
| 50 g | |
| 3655772 | |
| nicotinoyl chloride hydrochloride, pyridine-3-carbonyl chloride hydrochloride, nicotinoyl chloride, hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride, 3-pyridinecarbonyl chloride hydrochloride, nicotinic acid chloride hydrochloride, nicotinoylchloride hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride 1:1, nicotinyl chloride hydrochloride, nicotinoyl chloride hcl | |
| MSYBLBLAMDYKKZ-UHFFFAOYSA-N | |
| pyridine-3-carbonyl chloride;hydrochloride | |
| 88438 | |
| 98% |
Chemical Identifiers
| 20260-53-1 | |
| 178.012 | |
| MSYBLBLAMDYKKZ-UHFFFAOYSA-N | |
| 88438 | |
| C1=CC(=CN=C1)C(=O)Cl.Cl |
| C6H5Cl2NO | |
| MFCD00012820 | |
| nicotinoyl chloride hydrochloride, pyridine-3-carbonyl chloride hydrochloride, nicotinoyl chloride, hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride, 3-pyridinecarbonyl chloride hydrochloride, nicotinic acid chloride hydrochloride, nicotinoylchloride hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride 1:1, nicotinyl chloride hydrochloride, nicotinoyl chloride hcl | |
| pyridine-3-carbonyl chloride;hydrochloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 243-648-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only