missing translation for 'onlineSavingsMsg'
Learn More

Nicotine Solution NMR Reference Standard, 20 mg/mL in DMSO-d6 (99.9 atom D), MilliporeSigma™ Supelco™

Catalog No. 111016558
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016558 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016558 Supplier Merck Emd Millipore Supplier No. 6132151EA

Specifications

CAS 54-11-5
Concentration 20 mg/mL in DMSO- d 6 (99.9 atom% D)
Linear Formula C10H14N2
Type NMR Reference Standard
Quantity 1 Ea.
Formula Weight 162.23
Grade NMR Reference Standard
Concentration or Composition (by Analyte or Components) CFCl 3 , 0.4%