missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotine Solution NMR Reference Standard, 2 mg/mL in methanol-d4 (99.8 atom D), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 811-98-3 |
| Linear Formula | C10H14N2 |
| Concentration | 2 mg/mL in methanol-d 4 (99.8 atom% D) |
| Type | NMR Reference Standard |
| Quantity | 1 Ea. |
| Formula Weight | 162.23 |
| Grade | NMR Reference Standard |
| Concentration or Composition (by Analyte or Components) | TMS, 0.05% |