missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide N-Oxide 98.0+%, TCI America™

Catalog No. p-7133924
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06695G 5 g
N066925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06695G Supplier TCI America Supplier No. N06695G
Only null left

Chemical Identifiers

CAS 1986-81-8
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00006202
InChI Key USSFUVKEHXDAPM-UHFFFAOYSA-N
PubChem CID 72661
IUPAC Name 1-oxidopyridin-1-ium-3-carboxamide
SMILES C1=CC(=C[N+](=C1)[O-])C(=O)N

Specifications

Color White-Yellow
Quantity 5 g
Synonym nicotinamide n-oxide, nicotinamide 1-oxide, 1-oxynicotinamide, 3-pyridinecarboxamide, 1-oxide, 1-oxy-nicotinamide, niacinamide-n-oxide, nicotinamide, 1-oxide, 3-pyridinecarboxamide,1-oxide, 3-carbamoylpyridin-1-ium-1-olate, 3-aminocarbonyl pyridinium-1-olate
Formula Weight 138.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nicotinamide N-Oxide