accessibility menu, dialog, popup

UserName

H2NONHOHC

CAS RN 1986-81-8

IUPAC Name: 3-carbamoyl-1,4-dihydro-1λ²-pyridine-1,1-diium-4-id-1-olate
Synonyms: 3-carbamoyl-1,4-dihydro-1λ²-pyridine-1,1-diium-4-id-1-olate
Molecular Weight (g/mol): 139.13
Molecular Formula: C6H7N2O2
InChi Key: GNDQXIOMBYJZAE-UHFFFAOYNA-N
SMILES: NC(=O)C1=C[NH++]([O-])C=C[CH-]1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)