missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide N-oxide, 98%

Catalog No. aah6129222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6129222 100 g
AAH6129214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6129222 Supplier Thermo Scientific Chemicals Supplier No. H6129222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1986-81-8 | C6H6N2O2 | 138.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1986-81-8
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00006202
InChI Key USSFUVKEHXDAPM-UHFFFAOYSA-N
PubChem CID 72661
IUPAC Name 1-oxidopyridin-1-ium-3-carboxamide
SMILES C1=CC(=C[N+](=C1)[O-])C(=O)N

Specifications

Melting Point 291°C to 293°C (decomposition)
Quantity 100 g
Sensitivity Hygroscopic
Synonym nicotinamide n-oxide, nicotinamide 1-oxide, 1-oxynicotinamide, 3-pyridinecarboxamide, 1-oxide, 1-oxy-nicotinamide, niacinamide-n-oxide, nicotinamide, 1-oxide, 3-pyridinecarboxamide,1-oxide, 3-carbamoylpyridin-1-ium-1-olate, 3-aminocarbonyl pyridinium-1-olate
Formula Weight 138.12
Percent Purity 98%
Chemical Name or Material Nicotinamide N-oxide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-859-4
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only