missing translation for 'onlineSavingsMsg'
Learn More

Nicorandil 98.0+%, TCI America™

Catalog No. p-7133991
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0837100MG 100 mg
N08371G 1 g
N08375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0837100MG Supplier TCI America Supplier No. N0837100MG
Only null left

Chemical Identifiers

CAS 65141-46-0
Molecular Formula C8H9N3O4
Molecular Weight (g/mol) 211.18
MDL Number MFCD00186520
InChI Key LBHIOVVIQHSOQN-UHFFFAOYSA-N
Synonym nicorandil, ikorel, 2-nicotinamidoethyl nitrate, adancor, dancor, nicorandilum, sigmart, 2-pyridine-3-carbonylamino ethyl nitrate, n-2-hydroxyethyl nicotinamide nitrate, nicorandilum inn-latin
PubChem CID 47528
ChEBI CHEBI:31905
IUPAC Name 2-[(pyridin-3-yl)formamido]ethyl nitrate
SMILES [O-][N+](=O)OCCNC(=O)C1=CC=CN=C1

Specifications

Melting Point 90°C
Color White
Quantity 100 mg
Formula Weight 211.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nicorandil
RTECSNumber US4667600
TSCA No