missing translation for 'onlineSavingsMsg'
Learn More

Nicorandil, 98+%

Catalog No. p-7058192
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60879MD 250 mg
AAJ6087903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60879MD Supplier Thermo Scientific Chemicals Supplier No. J60879MD

A vasodilator that activates ATP-sensitive potassium channels | CAS: 65141-46-0 | C8H9N3O4 | 211.18 g/mol

A vasodilator that activates ATP-sensitive potassium channels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65141-46-0
Molecular Formula C8H9N3O4
Molecular Weight (g/mol) 211.18
MDL Number MFCD00186520
InChI Key LBHIOVVIQHSOQN-UHFFFAOYSA-N
Synonym nicorandil, ikorel, 2-nicotinamidoethyl nitrate, adancor, dancor, nicorandilum, sigmart, 2-pyridine-3-carbonylamino ethyl nitrate, n-2-hydroxyethyl nicotinamide nitrate, nicorandilum inn-latin
PubChem CID 47528
ChEBI CHEBI:31905
IUPAC Name 2-[(pyridin-3-yl)formamido]ethyl nitrate
SMILES [O-][N+](=O)OCCNC(=O)C1=CC=CN=C1

Specifications

Melting Point 92°C to 93°C
Color White
Quantity 250 mg
Merck Index 14,6521
Formula Weight 211.17
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Nicorandil
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
EINECSNumber 265-514-1
RTECSNumber US4667600
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only