missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel wire, 1.0mm (0.04in) dia, hard, 99.5% (metals basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 10 m |
| Odor | Odorless |
| Weight | ≈7.0g/m |
| Form | Wire |
| Percent Purity | 99.5% |
| Assay | (metals basis) |
| Chemical Name or Material | Nickel wire, 1.0mm (0.04 in.) dia., Hard, Temper: as drawn |
Safety and Handling
| Hazard Category | H317-H351-H372 |
| Hazard Statement | GHS H Statement H372-H351-H317 Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c |
| EINECSNumber | 231-111-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only