missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel wire, 1.0mm (0.04in) dia, annealed, 99.98% (metals basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 8.908g/cm3 at 20°C |
| Quantity | 2 m |
| Odor | Odorless |
| Form | Wire |
| Assay Percent Range | 99.98%, metal basis |
| Chemical Name or Material | Nickel |
Safety and Handling
| Hazard Category | H317-H351-H372 |
| Hazard Statement | Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). |
| Precautionary Statement | P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c |
| EINECSNumber | 231-111-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only