missing translation for 'onlineSavingsMsg'
Learn More

Nickel wire, 0.5mm (0.02in) dia, annealed, 99.98% (metals basis)

Form: Approx.1.75g/m, Temper: soft | CAS: 7440-02-0 | Ni | 58.69 g/mol

Supplier:  Thermo Scientific Chemicals 043132G9

 View more versions of this product

Catalog No. AA43132G9


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver to Gray
7440-02-0
25 m
99.98% (Metals basis)
Ni
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
[Ni]
58.69
CHEBI:28112
Nickel
Silver gray wire
Odorless
Wire
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
935

Chemical Identifiers

7440-02-0
58.69
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137 MFCD06798735
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351-H372

EINECSNumber : 231-111-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only