missing translation for 'onlineSavingsMsg'
Learn More

Nickel wire, 0.1mm (0.004in) dia, Puratronic™, 99.994% (metals basis)

Catalog No. p-4412849
Change view
Click to view available options
Quantity:
2 m
10 m
50 m
Catalog No. Quantity
AA10932G2 2 m
AA10932G6 10 m
AA10932H2 50 m
3 options

Catalog No. AA10932G2

Supplier: Thermo Scientific Chemicals 010932G2

Only null left

Form: approx. 0.07g/m | CAS: 7440-02-0 | Ni | 58.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Color Silver to Gray
Appearance Silver gray wire
Quantity 2 m
Odor Odorless
Form Wire
Assay Percent Range 99.994% (Metals basis)
Chemical Name or Material Nickel
Hazard Category H317-H351-H372
Precautionary Statement P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 231-111-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only