missing translation for 'onlineSavingsMsg'
Learn More

Nickel wire, 0.025mm (0.001in) dia, hard, 99.98% (metals basis)

Catalog No. AA40672BU
Change view
Click to view available options
Quantity:
25 cm
50 cm
1 m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40672BU 25 cm
AA40672BW 50 cm
AA40672G1 1 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40672BU Supplier Thermo Scientific Chemicals Supplier No. 040672BU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7440-02-0 | Ni | 58.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 25 cm
Odor Odorless
Weight ≈0.0044g/m
Form Wire
Percent Purity 99.98%
Assay (metals basis)
Chemical Name or Material Nickel wire, 0.025mm (0.001 in.) dia., Hard, Temper: as drawn
Hazard Category H317-H351-H372
Hazard Statement GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 231-111-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only