missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Sulfate, Hexahydrate, Crystal, Reagent, ACS, 98-102%, Spectrum™ Chemical
N1070, 10101-97-0, NiSO4·6H2O
Supplier: Spectrum Chemical Mfg Cor N1070500GM
Description
Spectrum™ Chemical Nickel Sulfate, Hexahydrate, Crystal, Reagent, ACS is an inorganic compound that appears as a blue colored salt. Nickel Sulfate is also considered highly soluble and commonly used for electroplating of nickel. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Specifications
| 10101-97-0 | |
| 500 g | |
| RRIWRJBSCGCBID-UHFFFAOYSA-L | |
| nickel(2+) hexahydrate sulfate | |
| 98 to 102% | |
| Poly Bottle |
| 1 | |
| H12NiO10S | |
| O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O | |
| 262.84 | |
| ACS |
Chemical Identifiers
| 10101-97-0 | |
| 262.84 | |
| nickel(2+) hexahydrate sulfate |
| H12NiO10S | |
| RRIWRJBSCGCBID-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O |