missing translation for 'onlineSavingsMsg'
Learn More

Nickel Sulfate, Hexahydrate, Crystal, Reagent, ACS, 98-102%, Spectrum™ Chemical

Catalog No. p-7157897
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604802 25 g Poly Bottle
18604803 125 g Poly Bottle
18604804 500 g Poly Bottle
18604806 2.5 kg Poly Pail
18604805 6 x 500 g Poly Bottle
18604807 12 kg Poly Pail
18604808 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18604802 Supplier Spectrum Chemical Mfg Cor Supplier No. N107025GM
Only null left

N1070, 10101-97-0, NiSO4·6H2O

Spectrum™ Chemical Nickel Sulfate, Hexahydrate, Crystal, Reagent, ACS is an inorganic compound that appears as a blue colored salt. Nickel Sulfate is also considered highly soluble and commonly used for electroplating of nickel. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.84
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
IUPAC Name nickel(2+) hexahydrate sulfate
SMILES O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98 to 102%
Grade ACS
Packaging Poly Bottle