missing translation for 'onlineSavingsMsg'
Learn More

Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3

CAS: 7440-02-0 | Ni | 58.69 g/mol

$46.44 - $144.87

Specifications

Chemical Name or Material Nickel standard solution
Name Note 1 mg/ml Ni in 2% HNO3
Packaging Plastic bottle
CAS 7440-02-0,13478-00-7,7697-37-2,7732-18-5
CAS Min % 1.9,96.9
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC196151000
View Documents
Thermo Scientific Chemicals
196151000
100 mL
Each for $46.44
Only null left
 
AC196155000
View Documents
Thermo Scientific Chemicals
196155000
500 mL
Each for $144.87
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
Specifications

Specifications

Nickel standard solution
Plastic bottle
1.9,96.9
1.0320g/mL
Crystals, Crystalline Powder and/or Chunks
990 to 1010 μg/mL Ni in 2 to 5% HNO3 (20°C)
Green
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
935
58.69
1 mg/ml Ni in 2% HNO3
7440-02-0,13478-00-7,7697-37-2,7732-18-5
2.1,97.1
1.032
Ni
MFCD00011137 MFCD06798735
Soluble in water
[Ni]
58.69
CHEBI:28112
AAS
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only