missing translation for 'onlineSavingsMsg'
Learn More

Nickel slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 42329QJ

 View more versions of this product

Catalog No. AA42329QJ


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel slug
7440-02-0
Odorless
Slug
Ni
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
[Ni]
58.69
CHEBI:28112
6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length
5 Slugs
≈3.58g/slug
(metals basis)
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
935
99.98%

Chemical Identifiers

7440-02-0
58.69
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137 MFCD06798735
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351-H372

EINECSNumber : 231-111-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only