missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Puratronic™, 99.995% (metals basis)
Form: approx. 0.22g/slug | CAS: 7440-02-0 | Ni | 58.69 g/mol
$140.51 - $1802.38
Chemical Identifiers
| CAS | 7440-02-0 |
|---|---|
| Molecular Formula | Ni |
| Molecular Weight (g/mol) | 58.69 |
| MDL Number | MFCD00011137 MFCD06798735 |
| InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
| Synonym | raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum |
| PubChem CID | 935 |
| ChEBI | CHEBI:28112 |
| IUPAC Name | nickel |
| SMILES | [Ni] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4233309
|
Thermo Scientific Chemicals
04233309 |
10 g |
Each for $140.51
|
|
|||||
|
AA4233318
|
Thermo Scientific Chemicals
04233318 |
50 g |
Each for $458.97
|
|
|||||
|
AA4233330
|
Thermo Scientific Chemicals
04233330 |
250 g |
Each for $1,802.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-02-0 | |
| 58.69 | |
| PXHVJJICTQNCMI-UHFFFAOYSA-N | |
| 935 | |
| nickel |
| Ni | |
| MFCD00011137 MFCD06798735 | |
| raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
| CHEBI:28112 | |
| [Ni] |
Specifications
| Silver to Gray | |
| Silver gray slug | |
| Odorless | |
| Slug | |
| MFCD00011137 MFCD06798735 | |
| PXHVJJICTQNCMI-UHFFFAOYSA-N | |
| nickel | |
| 935 | |
| Nickel |
| 7440-02-0 | |
| 10 g | |
| 99.995% (Metals basis) | |
| Ni | |
| raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
| [Ni] | |
| 58.69 | |
| CHEBI:28112 |
Safety and Handling
P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351-H372
EINECSNumber : 231-111-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only