Learn More
Nickel rod, 5mm (0.2in) dia, Puratronic™, 99.999% (metals basis)
CAS: 7440-02-0 | Ni | 58.69 g/mol
$529.59 - $529.59
Chemical Identifiers
| CAS | 7440-02-0 |
|---|---|
| Molecular Formula | Ni |
| Molecular Weight (g/mol) | 58.69 |
| MDL Number | MFCD00011137 MFCD06798735 |
| InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
| Synonym | raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum |
| PubChem CID | 935 |
| ChEBI | CHEBI:28112 |
| IUPAC Name | nickel |
| SMILES | [Ni] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-02-0 | |
| 58.69 | |
| PXHVJJICTQNCMI-UHFFFAOYSA-N | |
| 935 | |
| nickel |
| Ni | |
| MFCD00011137 MFCD06798735 | |
| raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
| CHEBI:28112 | |
| [Ni] |
Specifications
| 7440-02-0 | |
| Odorless | |
| Rod | |
| MFCD00011137 MFCD06798735 | |
| PXHVJJICTQNCMI-UHFFFAOYSA-N | |
| nickel | |
| 935 | |
| 99.999% | |
| Nickel rod, 5mm (0.2 in.) dia. |
| 50 mm | |
| ≈1.75g/cm | |
| Ni | |
| raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
| [Ni] | |
| 58.69 | |
| CHEBI:28112 | |
| Puratronic | |
| (metals basis) |
Safety and Handling
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351-H372
EINECSNumber : 231-111-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only